Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-23 p40 Antibody (B-P24), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB388304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388304 Supplier Novus Biologicals Supplier No. NBP314601AF488
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL-12/IL-23 p40
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-P24
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Host Species Mouse
Immunogen Recombinant human IL-12/IL-23 p40
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3593
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.