Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17C Antibody (311522), HRP, Novus Biologicals™
SDP

Catalog No. NB349077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349077 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349077 Supplier Novus Biologicals Supplier No. FAB23061H
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-17C Monoclonal antibody specifically detects IL-17C in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IL-17C
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 311522
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-17C, His15-Gln194, Accession # NP_665833
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 27189
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.