Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RA/IL-17R Antibody (SR1226), Novus Biologicals™
SDP

Catalog No. NB258397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258397 100 μg
NB260014 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258397 Supplier Novus Biologicals Supplier No. NBP322223100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IL-17RA/IL-17R
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1226
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Host Species Rabbit
Immunogen A synthesized peptide derived from human IL-17RA/IL-17R (Uniprot #: Q96F46)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 23765
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.