Learn More
Description
Specifications
Specifications
| Antigen | IL-17RA/IL-17R |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | SR1226 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human IL-17RA/IL-17R (Uniprot #: Q96F46) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
