Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Shrimp Alkaline Phosphatase

Catalog No. p-4955857 Shop All Cytiva Products
Click to view available options
Quantity:
1000 U
500 U
5000 U

Includes

10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0

Catalyzes removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins

  • Bacterial
  • Expressed in E.Coli
  • Inactivated by heating at 75°C for 5 minutes
  • Active in PCR buffers and restriction endonuclease buffers
  • Activity: 1U/μL
  • Purified to homogeneity
  • Free of all contaminating endonucleases, exonucleases and ribonucleases
TRUSTED_SUSTAINABILITY

Specifications

Purity Purified to homogeneity
Source Bacteria
Quantity 500 U
Product Type Alkaline Phosphatase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.