Learn More
Description
Specifications
Specifications
| Antigen | ILT2/CD85j/LILRB1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | GHI/75 |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7 |
| Gene Symbols | LILRB1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
