Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Image-iT™ Green Hypoxia Reagent

Click to view available options
Quantity:
1 Vial
5 Vials
Description
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.
Features of Image-iT Green Hypoxia Reagent include:
• Indicates hypoxia in live cells by fluorescing in low oxygen environments
• Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
• Easy to use—just add to cell culture media and image
Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.
Features of Image-iT Green Hypoxia Reagent include:
• Indicates hypoxia in live cells by fluorescing in low oxygen environments
• Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
• Easy to use—just add to cell culture media and image
Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.
Order Info
- Store at -5 to -30°C. Protect from light.
- Stable for at least 6 months after receiving.

Specifications
Specifications
Content And Storage | Store at -5 to -30°C. Protect from light. |
Product Type | Reagent |
Product Line | Image-iT |
Quantity | 1 Vial |
Reagent Type | Hypoxia Reagent |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction