Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin alpha 5/KPNA1/SRP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen Importin alpha 5/KPNA1/SRP1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15460420
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP15460420UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP154604
SDP
missing translation for 'viewDocuments'
Novus Biologicals
NBP154604
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Importin alpha 5/KPNA1/SRP1 Polyclonal specifically detects Importin alpha 5/KPNA1/SRP1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Importin alpha 5/KPNA1/SRP1
Polyclonal
Rabbit
P52294
3836
Synthetic peptides corresponding to KPNA1(karyopherin alpha 1 (importin alpha 5)) The peptide sequence was selected from the N terminal of KPNA1. Peptide sequence NMEMAPGGVITSDMIEMIFSKSPEQQLSATQKFRKLLSKEPNPPIDEVIS.
Primary
60 kDa
Western Blot, Immunohistochemistry
Unconjugated
RUO
importin subunit alpha-1, importin-alpha-S1, IPOA5, karyopherin alpha 1 (importin alpha 5), Karyopherin subunit alpha-1, NPI-1SRP1importin alpha 5, Nucleoprotein interactor 1, RCH2RAG cohort protein 2, recombination activating gene cohort 2, SRP1-beta
KPNA1
IgG
This product is specific to Subunit or Isoform: alpha-1.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.