Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Nucleoprotein Antibody, Novus Biologicals™
SDP

Catalog No. p-5067163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056570T 0.025 mg
NB10056570 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056570T Supplier Novus Biologicals Supplier No. NB10056570SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Influenza A H1N1 Nucleoprotein Polyclonal specifically detects Influenza A H1N1 Nucleoprotein in Human, Mouse, Influenza A Virus H1N1, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Influenza A H1N1 Nucleoprotein
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 24086762), Immunohistochemistry-Paraffin reported in scientific literature (PMID 16873262)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_001081025.1
Host Species Rabbit
Immunogen Recombinant Influenza A H1N1 Nucleoprotein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Test Specificity Influenza A virus (A/Puerto Rico/8/34/Mount Sinai (H1N1) segment 5) nuclear protein.
Target Species Human, Mouse, Influenza virus, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.