Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H7N7 Neuraminidase Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB350727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350727 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350727 Supplier Novus Biologicals Supplier No. NBP314650PCP
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A H7N7 Neuraminidase Polyclonal antibody specifically detects Influenza A H7N7 Neuraminidase in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Influenza A H7N7 Neuraminidase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Influenza A H7N7 Neuraminidase (Accession#: AAR11367.1; His36-Ser471).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.