Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ INSTA-Blot Human Tissues
SDP

Catalog No. NBP231378 Shop All Novus Biologicals Products
Click to view available options
Product Type:
Human Tissues Membrane

Applications: Western Blot

Recommended Storage

Store at room temperature

Specifications

For Use With (Application) Tissues are from donors with no known disease. Tissues are lysed in lysis buffer (10 mM Tris, pH 8.0, 130 mM NaCl, 1% Triton X-100, 10 mM NaF, 10 mM NaPi, 10 mM NaPPi) containing Protease Inhibitor Cocktail (PIC) and PMSF. Prior to SDS-PAGE the tissue lysate is resuspended with 4x Laemmli sample buffer with B-mercaptoethanol and denatured in boiling water. Use in Western blot reported in scientific literature (PMID 25615412)
Product Type Human Tissues Membrane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.