Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ INSTA-Blot Rat Tissues
SDP

Catalog No. NBP230112 Shop All Novus Biologicals Products
Click to view available options
Quantity:
1 Membrane

PVDF membrane containing denatured protein from cells or tissue lysates

  • Screening proteins from various rat tissues
  • Contains 11 rat tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
  • Prepared from resolution by SDS-PAGE at 125V for 1 hour
  • Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
  • Stained with amido black for visualization of proteins
Applications: 
Screening proteins from various rat tissues

Recommended Storage

Store at room temperature

Specifications

For Use With (Application) Screening proteins from various rat tissues
Quantity 1 Membrane
Material PVDF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.