Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 8 Antibody (CL7290), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Integrin beta 8 |
---|---|
Clone | CL7290 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Description
Integrin beta 8 Monoclonal specifically detects Integrin beta 8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Integrin beta 8 | |
Monoclonal | |
Mouse | |
integrin beta-8, integrin, beta 8 | |
ITGB8 | |
IgG1 | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL7290 | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
3696 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: AQHCVNSKGQVCSGRGTCVCGRCECTDPRSIGRFCEHCPTCYTACKENWNCMQCLHPHNLSQAILDQCKTSCALMEQQHY | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title