Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AccuPrime™ Pfx DNA Polymerase

AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis.

$626.00 - $2815.00

Specifications

Detection Method Primer-probe
GC-Rich PCR Performance Low
Polymerase AccuPrime Pfx DNA Polymerase
Reaction Speed Standard
Shipping Condition Wet or Dry Ice
View More Specs

Includes: AccuPrime Pfx DNA Polymerase (100μL, at 2.5U/μL) is provided with 10X AccuPrime reaction mix (1mL) that contains dNTPs and a separate vial (1mL) of 50mM MgSO4. Reagents are provided for 200 amplification reactions of 50μL each.

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
12-344-024
Encompass_Preferred
View Documents
Invitrogen™
12344024
200 Reactions
Each for $626.00
Only null left
Add to Cart
 
12-344-032
Encompass_Preferred
View Documents
Invitrogen™
12344032
1000 Reactions
Each for $2,815.00
Only null left
Add to Cart
 
Description

Description

AccuPrime Pfx DNA Polymerase is ideal for high-fidelity amplification of DNA fragments for downstream applications such as cloning and mutagenesis. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading (3'→5' exonuclease) activity. Platinum anti-Pfx DNA polymerase antibodies inhibit polymerase activity, providing hot-start capabilities for improved PCR specificity (see figure). Thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, increasing specificity, yield, and robustness over Platinum Pfx alone.

Benefits of AccuPrime Pfx DNA Polymerase

  • Greater than 26 times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 12 kb
  • Minimized PCR optimization

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • High-fidelity PCR
  • Hot-start PCR

Notes

  • AccuPrime Pfx DNA Polymerase is especially effective where very high levels of fidelity are required.
  • The 10X AccuPrime Pfx reaction mix contains MgSO4, dNTPs, and thermostable proteins that enhance specific primer-template hybridization during each PCR cycle.

Order Info

Approved for shipment on Wet or Dry Ice

Specifications

Specifications

Primer-probe
AccuPrime Pfx DNA Polymerase
Wet or Dry Ice
26X
Blunt
12 kb or less
Low
Standard
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Separate Components
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.