Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit

Catalog No. p-4922112
Encompass_Preferred
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions

Includes

100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X SYBR Green Reaction Mix; 1mL Magnesium Sulfate (50mM); 100μL ROX Reference Dye (25μM); 300μL 20X Bovine Serum Albumin (1mg/mL)

Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.

The SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.

  • Kit combines the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format
  • The SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection
  • SuperScript™ III RT for higher temperature cDNA synthesis for greater success with difficult RNA secondary structure
  • Platinum™ Taq DNA Polymerase with hot-start technology for improved specificity
  • LUX™ Fluorogenic Primers or dual-labeled fluorogenic probes for superior detection performance
  • SYBR™ Green I dye for easy and convenient detection
  • Validated performance on multiple real-time instrument platforms, including rotor-based systems
  • Store all components at -20°C (-80°C for long-term storage)
  • ROX Reference Dye must be stored in the dark

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 500 μL SuperScript™ III RT/Platinum™ Taq Mix
• 12.5 mL 2X SYBR™ Green Reaction Mix
• 2 × 1 mL Magnesium Sulfate (50 mM)
• 500 μL ROX Reference Dye (25 μM)
• 1.3 mL 20X Bovine Serum Albumin (1 mg/mL)

Store all components at -20°C (-80°C for long-term storage). ROX Reference Dye must be stored in the dark.
Detection Method SYBR
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
Product Line Platinum™, SYBR™, SuperScript™
Product Type One-Step qRT-PCR Kit
Quantity 500 reactions
Shipping Condition Dry Ice
Sufficient For 500 Reactions
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 500 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.