Learn More
Invitrogen™ Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System

Description
Includes
ThermoScript RT/Platinum Taq mix, and 2X ThermoScript reaction mix (buffer containing 0.4mM of each dNTP, 6mM MgSO4 ), vial of 50mM MgSO4
Order Info
Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice

Specifications
Specifications
Content And Storage | Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System contains ThermoScript™ RT/Platinum™ Taq mix, and 2X ThermoScript™ reaction mix (a buffer containing 0.4 mM of each dNTP, 6 mM MgSO4 ). A vial of 50 mM MgSO4 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored. |
Detection Method | Primer-probe |
PCR Method | 1-step RT-qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Line | Platinum™, ThermoScript™ |
Product Type | RT-PCR One-Step System |
Quantity | 100 reactions |
Shipping Condition | Dry Ice |
Sufficient For | 100 Reactions |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.