Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF5 Rabbit anti-Human, Mouse, Clone: 6L2B5, Novus Biologicals™
SDP

Catalog No. p-7246869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165783 20 μg
NB165782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165783 Supplier Novus Biologicals Supplier No. NBP31534320UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IRF5 Monoclonal antibody specifically detects IRF5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IRF5
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 6L2B5
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 6-151 of human IRF5 (Q13568). PVAPTPPRRVRLKPWLVAQVNSCQYPGLQWVNGEKKLFCIPWRHATRHGPSQDGDNTIFKAWAKETGKYTEGVDEADPAKWKANLRCALNKSRDFRLIYDGPRDMPPQPYKIYEVCSNGPAPTDSQPPEDYSFGAGEEEEEEEELQ
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3663
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.