Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISG20 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15721320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15721320 20 μL
NBP157213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15721320 Supplier Novus Biologicals Supplier No. NBP15721320UL

Rabbit Polyclonal Antibody

ISG20 Polyclonal specifically detects ISG20 in Human samples. It is validated for Western Blot.

Specifications

Antigen ISG20
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q96AZ6
Gene Alias CD25, Estrogen-regulated transcript 45 protein, HEM45EC 3.1.13.1, interferon stimulated exonuclease gene 20kDa, interferon stimulated gene (20kD), interferon-stimulated gene 20 kDa protein, Promyelocytic leukemia nuclear body-associated protein ISG20
Gene Symbols ISG20
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ISG20(interferon stimulated exonuclease gene 20kDa) The peptide sequence was selected from the middle region of ISG20. Peptide sequence TSTDRLLWREAKLDHCRRVSLRVLSERLLHKSIQNSLLGHSSVEDARATM.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3669
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.