Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Ether (Certified ACS), Fisher Chemical™

Catalog No. E1414 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E141-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E141-4
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-20-3 , 128-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Boiling Range 2 °C max., Inclusive of 67 °C
Melting Point -85°C
Density .7250g/cm³
Boiling Point 68°C
Flash Point −27°C
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2O
Quantity 4 L
UN Number 1159
Residue after Evaporation 0.01 % max.
Specific Gravity Inclusive Between 0.720
Stabilizer Inclusive Between 90ppm
Vapor Density 3.52g/mol
Vapor Pressure 150mmHg at 25°C
Formula Weight 102.18
Grade Certified
Physical Form Liquid
Chemical Name or Material Isopropyl Ether
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.