Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITF46 Antibody, Novus Biologicals™
SDP

Catalog No. NB405723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB405723 25 μL
NBP189910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB405723 Supplier Novus Biologicals Supplier No. NBP18991025UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

ITF46 Polyclonal specifically detects ITF46 in Human, Zebrafish samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ITF46
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias C11orf2UPF0360, C11orf60, chromosome 11 open reading frame 60, FLJ21827, intraflagellar transport 46 homolog (Chlamydomonas), intraflagellar transport protein 46 homolog, intraflagellar transport protein IFT46
Gene Symbols IFT46
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ETDSDSDDDDEEHGAPLEGAYDPADYEHLPVSAEIKELFQYISRYTPQLIDLDHKLKPFIPDFIPAVGDIDAFLKVPRPDGKPDNLGLLV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56912
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.