Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGB1BP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ITGB1BP3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15303320
![]() |
Novus Biologicals
NBP15303320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP153033
![]() |
Novus Biologicals
NBP153033 |
100 μL |
Each for $487.50
|
|
|||||
Description
ITGB1BP3 Polyclonal specifically detects ITGB1BP3 in Human samples. It is validated for Western Blot.Specifications
ITGB1BP3 | |
Polyclonal | |
Rabbit | |
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
EC 2.7.1.22, EC 2.7.1.n4, integrin beta 1 binding protein 3, Integrin beta-1-binding protein 3, MGC126624, MIBPNRK 2, Muscle integrin-binding protein, muscle-specific beta 1 integrin binding protein, nicotinamide riboside kinase 2, Nicotinic acid riboside kinase 2, NmR-K 2, NRK2RNK 2, Ribosylnicotinamide kinase 2, Ribosylnicotinic acid kinase 2 | |
NMRK2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q9NPI5 | |
27231 | |
Synthetic peptides corresponding to ITGB1BP3(integrin beta 1 binding protein 3) The peptide sequence was selected from the middle region of ITGB1BP3. Peptide sequence YKPLVDLYSRRYFLTVPYEECKWRRSTRNYTVPDPPGLFDGHVWPMYQKY. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title