Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 16259685
GREENER_CHOICE

Catalog No. 233350
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-335-0 50 mg
23-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-335-0 Supplier Tocris Bioscience™ Supplier No. 2333/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent, mGlu1-selective non-competitive antagonist

Sub-nanomolar potent, non-competitive mGlu1 antagonist (Ki = 0.34 nM). Inhibits glutamate-induced Ca2+ response at the human mGlu1 receptor with an IC50 value of 0.55 nM. Selective over mGlu5 (> 400-fold) and displays no activity at mGlu2, mGlu3, mGlu4, mGlu6, AMPA or NMDA receptors (IC50 > 10 μM). Centrally active following systemic administration.

Chemical Identifiers

CAS 409345-29-5
Molecular Formula C20H23NO3

Specifications

Quantity 50 mg
Formula Weight 325.41
Purity >98%
Molecular Weight (g/mol) 325.408
Chemical Name or Material JNJ 16259685
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.