Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JNJ 303

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent IKs blocker (IC50 = 64 nM). Displays no effects on other cardiac channels; IC50 values are 3.3, >10, 11.1 and 12.6 μM for INa, ICa, Ito and IKr currents respectively. Prolongs QT and causes unprovoked torsades de pointes (TdP).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 440.98 |
| Purity | >98% |
| Molecular Weight (g/mol) | 440.983 |
| Chemical Name or Material | JNJ 303 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction