Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Ab-2 Rabbit Polyclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-2, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$445.00 - $1099.00

Specifications

Antigen Kappa Light Chain Ab-2
Clone Polyclonal
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB333A1
View Documents
Epredia
RB333A1
Concentrated 500 μL
Each for $445.00
Only null left
Add to Cart
 
RB333A
View Documents
Epredia
RB333A
Concentrated 1 mL
Each for $1,099.00
Only null left
Add to Cart
 
RB333R1
View Documents
Epredia
RB333R1
Ready-to-Use 1 mL N/A N/A N/A
RB333RQ
View Documents
Epredia
RB333RQ
Ready-to-Use 12 mL N/A N/A N/A
Description

Description

Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey and Rat. Does not react with dog. Others not known.

Immunogen: Kappa light chains purified from human Bence Jones proteins

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Kappa Light Chain Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
IVD
kappa light chains purified from human Bence Jones proteins
Polyclonal
Polyclonal
Rabbit
Hematopoietic
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.