Learn More
Dynalon™ Kartell Bucket with Graduations and Spout
SCI_ED
Catalog No. s37257 Shop All Dynalab Corporation Products
Click to view available options
Diameter (English) Outer:
11.02 in.
Height (English):
11.81 in.
For measuring and transporting liquids or solids. Dynalon™ Kartell Bucket with Graduations and Spout are chemical resistant especially at room temperature.
  • Features steel bale handle with comfortable grip
  • Made of LDPE
  • Graduated internally in liters
  • Use up to 80˚C or well below freezing
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.