Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNJ1 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018505 0.1 mg
NBP130792 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018505 Supplier Novus Biologicals Supplier No. NBP1307920.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNJ1 Polyclonal specifically detects KCNJ1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen KCNJ1
Applications Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml
Formulation Sterilized water with No Preservative
Gene Accession No. P48048
Gene Alias ATP-regulated potassium channel ROM-K, ATP-sensitive inward rectifier potassium channel 1, inwardly rectifying K+ channel, Kir1.1, potassium inwardly-rectifying channel, subfamily J, member 1, ROMK1inwardly rectifying subfamily J member 1
Gene Symbols KCNJ1
Host Species Rabbit
Immunogen A synthetic peptide from the cytoplasmic domain of human KCNJ1 (ROMK1, Kir1.1) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3758
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse (92%), Rat (92%).
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.