Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNK9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | KCNK9 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KCNK9 Polyclonal specifically detects KCNK9 in Mouse samples. It is validated for Immunohistochemistry.Specifications
| KCNK9 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| 51305 | |
| IgG | |
| Unpurified | |
| Specific for KCNK9. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| KT3.2, potassium channel, subfamily K, member 9, TASK-3 | |
| KCNK9 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title