Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 13X, 8 to 12 mesh

Catalog No. AC197310050
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum

CAS: 63231-69-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Color Beige
Quantity 5 kg
Packaging Plastic drum
Bulk Density 600g/L min.
Residual Water 1.5% max.
Water Adsorption 23.5% min.
Physical Form Beads
Chemical Name or Material Molecular sieves 13X, 13X, 8 to 12 mesh
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.