Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Straight-Sided Serum Stoppers
SDP

For research and pharmaceutical packaging applications

$203.00 - $721.00

Specifications

Certifications/Compliance USP, EP, JP
Description Latex free
Color Gray
Product Type Plug Stopper
For Use With (Application) For research and pharmaceutical packaging applications
View More Specs

Products 3
Catalog Number Mfr. No. Material Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Material Diameter (Metric) Outer Price Quantity & Availability  
03-340-302
SDP
View Documents
DWK Life Sciences
73811T21
Chlorobutyl Rubber, PTFE 20 mm
Case of 100 for $266.50
Only null left
Add to Cart
 
03-340-303
SDP
View Documents
DWK Life Sciences
7381121
Chlorobutyl Rubber 20 mm
Case of 1000 for $721.00
Only null left
Add to Cart
 
03-340-300
SDP
View Documents
DWK Life Sciences
73811T13
Chlorobutyl Rubber, PTFE 13 mm
Case of 100 for $203.00
Only null left
Add to Cart
 
Description

Description

  • For use with 13 and 20mm serum-finished bottles and vials
  • Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
  • Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors or natural rubber latex
  • Applicable to aqueous solutions with a pH range of 2 to 10
  • PTFE facing improves chemical compatibility and minimizes surface interactions

Order Info

Durometer 50.

Specifications

Specifications

USP, EP, JP
Gray
For research and pharmaceutical packaging applications
Latex free
Plug Stopper
50 Durometer
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.