Learn More
Promega Kinase Selectivity Profiling System: CAMK-2, 8 x 50 reactions
Catalog No. PRV6924 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
8 x 50 Reactions
Catalog No. Quantity
PRV6924 8 x 50 Reactions
1 options
Catalog No. PRV6924 Supplier Promega Supplier No. V6924
Only null left
Add to Cart
Add to Cart
Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.
Specifications
Quantity 8 x 50 Reactions
Content And Storage <-65°C