Learn More
Promega Kinase Selectivity Profiling System: General Panel, 24 x 50 reactions
Catalog No. PRV6928 Shop All Promega Corporation Products
Click to view available options
Quantity:
24 x 50 Reactions
Catalog No. PRV6928
Supplier: Promega V6928
Only null left
Add to Cart
Add to Cart
Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.
  • Selectivity Screening of Library Compounds for Effect on Panels of Kinases
  • 24 kinase and substrate pairs organized in three 8−tube strips
  • Simple reaction assembly and automation−friendly protocol
  • Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Quantity 24 x 50 Reactions
Content And Storage <-65°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.