Learn More
Promega Kinase Selectivity Profiling System: Other-2, 8 x 50 reactions
Shop All Promega Corporation ProductsDescription
- Selectivity Screening of Library Compounds for Effect on a Panel of Kinases
- Kinase and substrate pairs organized in an easy−to−use 8−tube strip format
- Simple reaction assembly and automation−friendly protocol
- Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Specifications
| Quantity | 8 x 50 Reactions |
| Content And Storage | <-65°C |