Learn More
Promega Kinase Selectivity Profiling System: Other/CK-1, 8 x 50 reactions
Catalog No. PRV6918 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
8 x 50 Reactions
Catalog No. Quantity
PRV6918 8 x 50 Reactions
1 options
Catalog No. PRV6918 Supplier Promega Supplier No. V6918
Only null left
Add to Cart
Add to Cart
Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.
  • Selectivity Screening of Library Compounds for Effect on a Panel of Kinases
  • Kinase and substrate pairs organized in an easy−to−use 8−tube strip format
  • Simple reaction assembly and automation−friendly protocol
  • Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Quantity 8 x 50 Reactions
Content And Storage <-65°C