Learn More
Promega Kinase Selectivity Profiling System: Other/CK-1 + ADP-Glo™ Assay
Shop All Promega Corporation ProductsDescription
- Selectivity Screening of Library Compounds for Effect on a Panel of Kinases
- Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
- Simple reaction assembly and automation-friendly protocol
- Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Specifications
| Quantity | 8 x 50 Reactions |
| Content And Storage | Components at different temperatures |
| Format | 384-well Plate |
| For Use With (Application) |
|
| Kit Contents | Cat.# V6918 Kinase Selectivity Profiling System: Other/CK-1, which includes:
|
| Product Type | Kinase Selectivity Profiling System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.