Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Kinase Selectivity Profiling System: STE-1 + ADP-Glo™ Assay
Shop All Promega Corporation Products
Click to view available options
Quantity:
8 x 50 Reactions
Description
- Selectivity Screening of Library Compounds for Effect on STE and TKL Kinases
- Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
- Simple reaction assembly and automation-friendly protocol
- Streamilines profiling of hits and leads for potency against families of related kinases
Specifications
Specifications
Quantity | 8 x 50 Reactions |
Content And Storage | Components at different temperatures |
Format | 384-well Plate |
For Use With (Application) |
|
Kit Contents | Cat.# V6916 Kinase Selectivity Profiling System: STE-1, which includes:
|
Product Type | Kinase Selectivity Profiling System |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction