Learn More
Kinematica HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
$729.50 - $2180.00
Catalog Number | Mfr. No. | Model | Dimensions (Dia. x H) | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Dimensions (Dia. x H) | Capacity (Metric) | Price | Quantity & Availability | |||
08-451-402
|
Kinematica HMC Europe
51009 |
Wire mesh basket | 338 x 235 mm | 80 L (Total), 50 L (Nominal) |
Each for $878.00
|
|
|||
08-451-403
|
Kinematica HMC Europe
51019 |
Wire mesh basket | 345 x 310 mm | 80 L (Total), 50 L (Nominal) |
Each for $1,457.00
|
|
|||
08-451-404
|
Kinematica HMC Europe
51010 |
Wire mesh basket with bottom | 345 x 220 mm | 80 L (Total), 50 L (Nominal) |
Each for $1,739.00
|
|
|||
08-451-405
|
Kinematica HMC Europe
51005 |
Long basket with closed bottom | 345 x 455 mm | 80 L (Total), 50 L (Nominal) |
Each for $2,180.00
|
|
|||
08-451-414
|
Kinematica HMC Europe
50006 |
Stackable basket | 225 x 250 mm | 25 L |
Each for $729.50
|
|
|||
08-451-417
|
Kinematica HMC Europe
50009 |
Stackable basket | 390 x 380 mm | 110 L |
Each for $1,084.00
|
|
|||
08-451-423
|
Kinematica HMC Europe
50027 |
With collecting pan | 280 x 260 mm | 25 L |
Each for $1,239.00
|
|
|||
08-451-424
|
Kinematica HMC Europe
50028 |
With collecting pan | 280 x 225 mm | 50 L |
Each for $1,611.00
|
|
|||
08-451-425
|
Kinematica HMC Europe
50029 |
With collecting pan | 390 x 290 mm | 85 L |
Each for $1,740.00
|
|
|||
08-451-426
|
Kinematica HMC Europe
50030 |
With collecting pan | 390 x 380 mm | 110 L |
Each for $1,980.00
|
|
|||
Description
For use with the HMC-HG and HMC-HV autoclaves.Specifications
Stainless Steel |