Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir3.3 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094665 100 μL
NB094666 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094665 Supplier Novus Biologicals Supplier No. NBP302950100UL

Rabbit Polyclonal Antibody

Kir3.3 Polyclonal specifically detects Kir3.3 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Kir3.3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias G protein-activated inward rectifier potassium channel 3, G protein-coupled inward rectifier potassium channel, GIRK-3, GIRK3KIR3.3, Inward rectifier K(+) channel Kir3.3, inwardly rectifier K+ channel KIR3.3, Kir3.3, Potassium channel, inwardly rectifying subfamily J member 9, potassium inwardly-rectifying channel, subfamily J, member 9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Kir3.3 (NP_004974.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3765
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.