Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KML 29
GREENER_CHOICE

Catalog No. p-4835940
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent and selective MAGL inhibitor

Highly selective and potent monoacylglycerol lipase (MAGL) inhibitor. Exhibits potent inhibition of human, mouse and rat MAGL (IC50 values are 5.9, 15 and 43 nM, respectively). Exhibits no detectable inhibition of FAAH (IC50 > 50000 nM). Potently and selectively blocks hydrolysis of 2-arachidonoylglycerol (2-AG) in mice (IC50 = 2.5 nM and >50 μM for 2-AG and AEA respectively).

Chemical Identifiers

CAS 1380424-42-9
Molecular Formula C24H21F6NO7

Specifications

Quantity 10 mg
Formula Weight 549.42
Molecular Weight (g/mol) 549.422
Chemical Name or Material KML 29
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.