Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv7.3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen Kv7.3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17410220
SDP
View Documents
Novus Biologicals
NBP17410220UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP174102
SDP
View Documents
Novus Biologicals
NBP174102
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Kv7.3 Polyclonal specifically detects Kv7.3 in Mouse, Hamster samples. It is validated for Western Blot.
Specifications

Specifications

Kv7.3
Polyclonal
Rabbit
Q8K3F6
3786
IgG
Western Blot
Unconjugated
RUO
BFNC2, EBN2, KQT-like 3, KV7.3, Potassium channel subunit alpha KvLQT3, potassium channel, voltage-gated, subfamily Q, member 3, potassium voltage-gated channel subfamily KQT member 3, potassium voltage-gated channel, KQT-like subfamily, member 3, Voltage-gated potassium channel subunit Kv7.3
KCNQ3
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.