Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals

Catalog No. p-4813502
Click to view available options
Quantity:
25 g
100 g

Catalog No. AC449480250

Supplier: Thermo Scientific Chemicals 449480250

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
Synonym ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf
PubChem CID 54680660
IUPAC Name [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

Melting Point 111°C to 115°C
Boiling Point 250°C
Ignition Residue 0.1% max
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 414.53
Chemical Name or Material L(+)-Ascorbyl palmitate
EINECSNumber 205-305-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.