Learn More
Stoelting™ Lab Standard™ Stereotaxic Instrument for Rats
SDP
Classic Lab Standard™ includes a 100 micron 3-axes, left-hand manipulator arm, rat adaptor, traditional (18°) ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized.
$6590.00 - $56450.00
Specifications
For Use With (Application) Neuroscience
Animal Rat
View More Specs
Products 18
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-133
SDP
View Documents
Stoelting™
51600
Lab Standard
Each for $8,380.00
Only null left
Add to Cart
10-000-134
SDP
View Documents
Stoelting™
51601
Lab Standard, Base only
Each for $6,590.00
Only null left
Add to Cart
10-000-136
SDP
View Documents
Stoelting™
51603
Dual Lab Standard
Each for $12,190.00
Only null left
Add to Cart
10-000-173
SDP
View Documents
Stoelting™
51650
Lab Standard, non-rupture ear bars
Each for $8,240.00
Only null left
Add to Cart
10-000-174
SDP
View Documents
Stoelting™
51653
Dual Lab Std ,non-rupture ear bars
Each for $11,840.00
Only null left
Add to Cart
10-000-188
SDP
View Documents
Stoelting™
51700
Motorized Lab Standard, 18 deg.
Each for $25,900.00
Only null left
Add to Cart
10-000-206
SDP
View Documents
Stoelting™
51750
Motorized Lab Standard, 45 deg.
Each for $23,320.00
Only null left
Add to Cart
10-000-207
SDP
View Documents
Stoelting™
51753
Motorized Dual Lab Standard
Each for $56,450.00
Only null left
Add to Cart
10-000-230
SDP
View Documents
Stoelting™
51900
Digital Lab Standard
Each for $11,840.00
Only null left
Add to Cart
10-000-231
SDP
View Documents
Stoelting™
51903
Digital Dual Lab Standard
Each for $19,000.00
Only null left
Add to Cart
10-000-238
SDP
View Documents
Stoelting™
51950
Digital Lab Std,nonrupture ear bars
Each for $11,840.00
Only null left
Add to Cart
10-000-239
SDP
View Documents
Stoelting™
51953
Digital Dual Lab Std/nonrup.ear bar
Each for $19,000.00
Only null left
Add to Cart
10-000-394
SDP
View Documents
Stoelting™
51600U
Ultra Precise Lab Standard
Each for $10,050.00
Only null left
Add to Cart
10-000-396
SDP
View Documents
Stoelting™
51603U
Ultra Precise Dual Lab Standard
Each for $15,430.00
Only null left
Add to Cart
10-000-404
SDP
View Documents
Stoelting™
51650U
Ultra Precise, non-rupture ear bars
Each for $10,050.00
Only null left
Add to Cart
10-000-405
SDP
View Documents
Stoelting™
51653U
Ultra Precise Dual, nonrup. ear bar
Each for $15,430.00
Only null left
Add to Cart
10-000-418
SDP
View Documents
Stoelting™
51900U
Ultra Precise Digital
Each for $13,620.00
Only null left
Add to Cart
10-000-420
SDP
View Documents
Stoelting™
51903U
Ultra Precise Dual Digital
Each for $22,590.00
Only null left
Add to Cart
Description
Description
  • Large, easy to read vernier scales Scales are laser engraved — accurate to 100 microns
  • Triple lead screws for fast positioning 80 mm of vertical, lateral and anterior-posterior travel
  • Absolute lock at 90° (vertical)
  • Brass bushings in manipulator arm permit electrical grounding
  • Accessories available for use with a wide variety of small animals
  • Adaptors available for use with mice, neonatal rats, birds and other species
  • Gas anesthesia compatible
  • Easily cleaned durable surface
  • Warmer ready base compatible with Stoelting Rodent Warmer System
  • Versatile Positioning
  • Manipulator arm controls medial-lateral and ventral-dorsal using exclusive triple lead screws for faster positioning, and anterior-posterior movement via dovetail slide movement, with 80mm of travel possible in each direction.
  • Universal joint allows the researcher to change the angle of the probe up to 90° in either the anterior-posterior or mediallateral planes.
  • Improved locking mechanism on the Lab Standard™ will hold any angle position without slipping and provides an absolute lock at 90° vertical.
  • Easily-Read Scales
    • Oriented to be easily read from the open end of the U
    • Numerals are large and easy to read.
    • Scale lines are laser engraved, to allow the finest possible permanent marking of scales on all 3 axes.
    • Precise alignment with facing vernier scales gives accurate resolution to 100 microns.
  • Smooth, Consistent Movements - Triple-lead screws allow smooth consistent movements with the fastest positioning possible.
  • Warmer ready base
  • Base Plates are designed to be integrated with the Stoelting Rodent Warmer System (sold separately) and are used to maintain body temperature throughout surgical procedures.
  • Thermal heating pads are embedded into base plates to avoid cross contamination with easy clean-up and maintain a sterile environment during stereotaxic surgery.
  • Can be used as a general warming system or with a rectal thermal probe (sold separately) for homeothermic regulation.
Specifications
Specifications
Neuroscience
Rat
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.