Learn More
Epredia™ Lab Vision™ CD8 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD8 Ab-1, Mouse Monoclonal Antibody.
$765.00
Specifications
Antigen | CD8 Ab-1 |
---|---|
Clone | C8/144B |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS457R7
|
Epredia™
MS457R7 |
Prediluted | 7 mL |
Each for $765.00
|
|
||||
MS457S0
|
Epredia™
MS457S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS457S1
|
Epredia™
MS457S1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
MS457S
|
Epredia™
MS457S |
Concentrated | 1 mL | N/A | N/A | N/A | |||
MS457R1
|
Epredia™
MS457R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
CD8 molecule consists of two chains, termed α and Β chain, which are expressed as a disulphide-α β heterdimer or as an alpha α homodimer on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8+ Tcells expresses CD8 as αβheterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not Β chain).Host Species: Mouse
Clone: C8/144B
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: C-terminal
Immunogen: A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of α-chain of human CD8 molecule
Molecular Weight: 32 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
1. Mason DY, et. al. Journal of Clinical Pathology, 1992, 45(12):1084-8.
Specifications
CD8 Ab-1 | |
Monoclonal | |
Mouse | |
T-Cell | |
A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of alpha chain of human CD8 molecule |
C8/144B | |
Unconjugated | |
IVD | |
Human | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.