Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1, Mouse Monoclonal Antibody.

$525.00 - $933.00

Specifications

Antigen Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Clone T9; same as 3E7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS314S1
View Documents
Epredia™
MS314S1
Concentrated 500 μL
Each for $596.00
Only null left
Add to Cart
 
MS314RQ
View Documents
Epredia™
MS314RQ
Ready-to-Use 12 mL
Each for $525.00
Only null left
Add to Cart
 
MS314S
View Documents
Epredia™
MS314S
Supernatant 1 mL
Each for $933.00
Only null left
Add to Cart
 
MS314S0
View Documents
Epredia™
MS314S0
Concentrated 100 μL N/A N/A N/A
MS314R1
View Documents
Epredia™
MS314R1
Prediluted 1 mL N/A N/A N/A
Description

Description

Hepatitis B virus causes type B hepatitis and it belongs to hepadnavirus family. It is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Mouse

Clone: T9; same as 3E7

Isotype: IgG1/λ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified HBVsAg, subtype ayw3

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
ASR
Purified HbsAg, subtype ayw3
T9; same as 3E7
Monoclonal
Mouse
Infectious Disease
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.