Learn More
Epredia™ Lab Vision™ Hepatocyte Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatocyte Ab-1, Mouse Monoclonal Antibody.
$196.00 - $1042.00
Specifications
| Antigen | Hepatocyte Ab-1 |
|---|---|
| Clone | OCH1E5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
MS1810S0
|
Epredia™
MS1810S0 |
Concentrated | 100 μL |
Each for $196.00
|
|
||||
|
MS1810S1
|
Epredia™
MS1810S1 |
Concentrated | 500 μL |
Each for $560.50
|
|
||||
|
MS1810S
|
Epredia™
MS1810S |
Concentrated | 1 mL |
Each for $1,042.00
|
|
||||
|
MS1810R7
|
Epredia™
MS1810R7 |
Prediluted | 7 mL |
Each for $523.50
|
|
||||
|
MS1810R1
|
Epredia™
MS1810R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
|
MS1810RQ
|
Epredia™
MS1810RQ |
Ready-to-Use | 12 mL | N/A | N/A | N/A | |||
Description
Host Species: Mouse
Clone: OCH1E5
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Formalin-fixed failed human allograft liver that was mechanically disrupted
Positive Control: Liver CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Hepatocyte Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Formalin fixed failed human allograft liver that was mechanically disrupted |
| OCH1E5 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.