Learn More
Epredia™ Lab Vision™ Mesothelin Ab-1 Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelin Ab-1, Mouse Monoclonal Antibody.
$717.00 - $1183.00
Specifications
Antigen | Mesothelin Ab-1 |
---|---|
Clone | 5B2 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1320S1
|
Epredia™
MS1320S1 |
Concentrated | 500 μL |
Each for $717.00
|
|
||||
MS1320S
|
Epredia™
MS1320S |
Concentrated | 1 mL |
Each for $1,183.00
|
|
||||
MS1320R1
|
Epredia™
MS1320R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.Host Species: Mouse
Clone: 5B2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Molecular Weight: 40kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mesothelin Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG1 |
5B2 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.