Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ p53 Rabbit Monoclonal Antibody (Clone SP5)

Employ part of a comprehensive antibody portfolio with the Epredia™ p53, Rabbit Monoclonal Antibody (Clone SP5). Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Specifications

Antigen p53
Clone SP5
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RM9105S0
View Documents
Epredia™
RM9105S0
Concentrated 100 μL N/A N/A N/A
RM9105S1
View Documents
Epredia™
RM9105S1
Concentrated 500 μL N/A N/A N/A
RM9105R7
View Documents
Epredia™
RM9105R7
Prediluted 7 mL N/A N/A N/A
RM9105RQ
View Documents
Epredia™
RM9105RQ
Ready-to-Use 12 mL N/A N/A N/A
Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Rabbit

Clone: SP5

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant human full-length wild type p53 protein

Molecular Weight: 53kDa

Positive Control: MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

p53
Western Blot
Unconjugated
IVD
Recombinant human full-length wild type p53 protein
SP5
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.