Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pds1 Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Pds1 Ab-1, Mouse Monoclonal Antibody.
Specifications
Antigen | Pds1 Ab-1 |
---|---|
Clone | DCS-280 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1511P0
|
Epredia
MS1511P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1511P1ABX
|
Epredia
MS1511P1ABX |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1511PABX
|
Epredia
MS1511PABX |
Concentrated | 200 μL | N/A | N/A | N/A | |||
MS1511P1
|
Epredia
MS1511P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
MS1511P
|
Epredia
MS1511P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
MS1511R1
|
Epredia
MS1511R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
MS1511R7
|
Epredia
MS1511R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Pds1 is an anaphase inhibitor and plays important role in DNA damage and spindle check point pathways. Pds1 inhibits sister chromatid separation by binding and inhibiting Esp1, cystein protease that causes cleavage of the cohesin Scc1 that binds the sister chromatids together. Pds1 is responsible for targeting of Esp1 to nucleus and its binding to the spindle. Degradation of Pds1 occurs shortly before anaphase, which liberates Esp1 and is a prerequisite for anaphase entry. Pds1 is targeted for degradation by ubiquitination mediated by cyclosome/anaphase-promoting complex (APC) functioning as a ubiquitin ligase. In response to DNA damage Chk1 phosphorylates Pds1 to stabilize it against the APC mediated destruction, hence preventing the entry of such a cell into anaphase.Host Species: Mouse
Clone: DCS-280
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human recombinant full-length Pds1 protein
Molecular Weight: 42kDa
Positive Control: HeLa cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Pds1 Ab-1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
DCS-280 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Human recombinant full length Pds1 protein | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title