Learn More
Epredia™ Lab Vision™ Perforin Ab-2, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states Epredia™ Perforin Ab-2, Mouse Monoclonal Antibody.
$229.00 - $703.00
Specifications
| Antigen | Perforin Ab-2 |
|---|---|
| Clone | 5B10 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Form | Price | Quantity & Availability | ||||
|
MS1834S0
|
Epredia™
MS1834S0 |
100 μL | Concentrated |
Each for $229.00
|
|
||||
|
MS1834S1
|
Epredia™
MS1834S1 |
500 μL | Concentrated |
Each for $703.00
|
|
||||
|
MS1834R7
|
Epredia™
MS1834R7 |
7 mL | Prediluted |
Each for $590.00
|
|
||||
|
MS1834S
|
Epredia™
MS1834S |
1 mL | Concentrated | N/A | N/A | N/A | |||
|
MS1834R1
|
Epredia™
MS1834R1 |
1 mL | Prediluted | N/A | N/A | N/A | |||
Description
Perforin is a potent cytolytic pore-forming protein. It is a specific marker of functionally active cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Perforin is a key effector mechanisms in T cell-mediated cytotoxicity. It mediates cytolysis of target cells by membrane damage and apoptosis.Host Species: Mouse
Clone: 5B10
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal region of human perforin
Molecular Weight: 70kDa
Positive Control: Lymphoma
Cellular Localization: Granular and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
1. Lichtenheld M G, et al. (1988) Nature, 335:448-451.
2. Hameed A, et. al. American Journal of Pathology, 1992, 140(5):1025-30.
Specifications
| Perforin Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG1 |
| 5B10 | |
| Monoclonal | |
| Mouse | |
| T-Cell | |
| Recombinant protein corresponding to C-terminal region of human perforin | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.