Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Vimentin Rabbit Monoclonal Antibody

Vimentin, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Specifications

Antigen Vimentin
Clone SP20
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RM9120S0
View Documents
Epredia™
RM9120S0
Concentrated 100 μL N/A N/A N/A
RM9120S1
View Documents
Epredia™
RM9120S1
Concentrated 500 μL N/A N/A N/A
RM9120S
View Documents
Epredia™
RM9120S
Concentrated 1 mL N/A N/A N/A
RM9120R1
View Documents
Epredia™
RM9120R1
Prediluted 1 mL N/A N/A N/A
Description

Description

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Rabbit

Clone: SP20

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein encoding human vimentin

Molecular Weight: 57-60kDa

Positive Control: Sarcoma and melanoma

Cellular Localization: Cytoplasmic Clone SP20 is excellent for staining of formalin/paraffin with no special pretreatment.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Vimentin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG
SP20
Monoclonal
Rabbit
Cytoskeletal Protein
Recombinant protein encoding human vimentin
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.