Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Evidence Drying Cabinet: 5 ft.W
SDP

Catalog No. p-4892150 Shop All Labconco Corporation Products
Click to view available options
Includes:
UV light
Washdown system with pump
Plug Type:
China/Australia
North America
Schuko
UK
US
Voltage:
110 V
220 V

Provide a compact, space-saving method of securely drying evidence in your lab with the Labconco Protector Evidence Drying Cabinet. This durable, stainless steel-lined cabinet provides a secure ventilated area to dry evidence such as clothing and bedding and its HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors.

A built-in blower provides airflow through the cabinet interior to speed drying. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA and carbon filters and is exhausted back to the room through the top of the cabinet.

  • Durable, stainless steel-lined construction
  • HEPA and carbon-filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors
  • Ultraviolet light provides secondary disinfection of interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing
  • Lockable cabinet maintains the chain of custody
  • UV security lock limits access to the ultraviolet light switch
  • Safety glass door with magnetic seal maximizes visibility into the cabinet while withstanding exposure to ultraviolet light
  • Inlet roughing prefilter traps dust and other large particles in the air entering the cabinet
  • Storage area below cabinet houses drain bucket and holds miscellaneous cleaning supplies
  • Leak-tight brushed stainless-steel interior cleans easily
  • Three-position switch activates fan or ultraviolet light (only when UV security lock key is in ON position)
  • Spacious interior measures 26.5D x 32.25W x 56 in.H (81.9 x 67.3 x 142.2cm)
  • Stainless-steel hanging rod accommodates clothing, bedding and other tall or bulky items; three wire mesh shelves
  • Exhaust roughing prefilter traps particulates in the exhaust air to prolong the life of the HEPA filter
  • HEPA filter, 99.99% efficient on particles 0.3 micron, traps airborne particulates including biohazardous aerosols, as air is exhausted from cabinet interior
  • Large 14-quart plastic bucket, located in the storage area below the cabinet, collects liquids from the drain in the cabinet's floor to aid clean up

Compliance

ETL listed (Models 3400000 and 3400001). CE conformity marking (Models 3400002, 3400003 and 3400004).

Specifications

Product Type Evidence Drying Cabinet
No. of Doors 2
No. of Drawers None
Color Light Gray
No. of Shelves 3
Voltage 220 V
Width (English) Cabinet 60 in.
Plug Type US
Weight (English) 675 lb.
Width (English) 60 in.
Weight (Metric) 306 kg
Includes UV light
Height (Metric) Storage Area 30.48 cm
Hertz 50/60Hz
Options Available UV Light
Height (Metric) 200.7 cm
Height (Metric) Interior 30.48 cm
Width (Metric) Storage Area 91.44 cm
Height (English) 79 in.
Width (English) Storage Area 36 in.
Width (Metric) Cabinet 152.4cm
Width (Metric) Shelf 20.3 cm
With Casters Yes
Height (English) Storage Area 12 in.
Width (English) Shelf 8 in.
Width (Metric) 152.4 cm
Filter Type HEPA and Carbon filter
Cabinet Type Evidence drying lockable cabinet
Door Type Safety glass door with magnetic seal
Depth (Metric) Shelf 48.26 cm
Depth (Metric) 71.1 cm
Amperage 3 A
Depth (English) Interior 26.5 in.
Depth (English) Shelf 19 in.
Depth (English) 28 in.
Depth (Metric) Interior 67.31 cm
Material Stainless Steel
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.